Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, 乳鉢、乳棒なし

VWR®, 乳鉢、乳棒なし

20 ml, 32 mm, 50 mm
Catalog # 410-0108
Supplier:   VWR Collection
VWR®, 乳鉢、乳棒なし
Catalog # 410-0108
Supplier:  VWR Collection
VWR®, 乳鉢、乳棒なし
VWR®, 乳鉢、乳棒なし

20 ml, 32 mm, 50 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Porcelain, produced according to DIN 12906.

  • With spout
  • Glazed outside, grinding surface unglazed
  • Highly resistant to acids and alkalis
Specifications
  • 内径:
    50 mm
  • 容量:
    20 ml
  • 高さ:
    32 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.