VWR®, 乳棒
36 mm, 150 mm
Catalog # 410-0121
Supplier: VWR Collection
Jump to:
Product Details & Documents
Porcelain.
- Grinding surface unglazed
- Glazed handle
: Produced according to DIN 12906.
Documents
410-0122
Images 360
Specifications
- Head Ø:36 mm
- 長さ:150 mm