Orders LinkContactUsLinkComponent
VWR®, 乳棒
VWR®, 乳棒
36 mm, 150 mm
Catalog # 410-0121
Supplier:   VWR Collection
VWR®, 乳棒
Catalog # 410-0121
Supplier:  VWR Collection
VWR®, 乳棒
VWR®, 乳棒
36 mm, 150 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Porcelain.
  • Grinding surface unglazed
  • Glazed handle
 : Produced according to DIN 12906.
Documents
Specifications
  • Head Ø:
    36 mm
  • 長さ:
    150 mm