VWR®, 乳棒
42 mm, 175 mm
410-0122
Porcelain.
- Grinding surface unglazed
- Glazed handle
Certifications: Produced according to DIN 12906.
Documents
410-0122
Images 360
- Head Ø:42 mm
- 長さ:175 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



