VWR®, 試験管ラック、PP
Blue, 72 (6×12), 13 mm, 103,5×202×60 mm
Catalog # 212-0431
Supplier: VWR Collection
Jump to:
Product Details & Documents
PP.
- Single-piece moulded
- Good heat and chemical resistance
- Three tiers with contents always clearly visible
- Large labelling areas on the side surfaces
Documents
Specifications
- For tube Ø:13 mm
- 穴の数:72 (6×12)
- Colour:Blue
- Seq:01
- W×D×H:103,5×202×60 mm