Order Entry
Japan
Orders LinkContactUsLinkComponent
 

VWR®, 試験管ラック、PP

White, 72 (6×12), 13 mm, 103,5×202×60 mm
Catalog # 212-0434
Supplier:   VWR Collection
VWR®, 試験管ラック、PP
Catalog # 212-0434
Supplier:  VWR Collection
VWR®, 試験管ラック、PP
VWR®, 試験管ラック、PP

White, 72 (6×12), 13 mm, 103,5×202×60 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

PP.

  • Single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
Specifications
  • For tube Ø:
    13 mm
  • 穴の数:
    72 (6×12)
  • Colour:
    White
  • Seq:
    09
  • W×D×H:
    103,5×202×60 mm