VWR®, 試験管ラック
White, 9 (3×3), 30 mm, 110,6×110,6×84,2 mm
Catalog # 212-0399
Supplier: VWR Collection
Jump to:
Product Details & Documents
Nylon, glass fibre reinforced.
- Racks are single-piece moulded
- Good heat and chemical resistance
- Three tiers with contents always clearly visible
- Large labelling areas on the side surfaces
Documents
Specifications
- For tube Ø:30 mm
- 穴の数:9 (3×3)
- Colour:White
- Seq:04
- W×D×H:110,6×110,6×84,2 mm