Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, 試験管ラック

VWR®, 試験管ラック

Yellow, 72 (6×12), 13 mm, 103,5×202,5×60 mm
Catalog # 212-0406
Supplier:   VWR Collection
VWR®, 試験管ラック
Catalog # 212-0406
Supplier:  VWR Collection
VWR®, 試験管ラック
VWR®, 試験管ラック

Yellow, 72 (6×12), 13 mm, 103,5×202,5×60 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Nylon, glass fibre reinforced.

  • Racks are single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
Specifications
  • For tube Ø:
    13 mm
  • 穴の数:
    72 (6×12)
  • Colour:
    Yellow
  • Seq:
    54
  • W×D×H:
    103,5×202,5×60 mm