酵母エキス、VWR
Yeast extract, 10 kg
Catalog # J850-10KG
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Yeast extract is the water soluble portion of autolysed yeast containing a source of Vitamin B complex.
- High purity extract
- Used for supplementing media
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:10 kg
- 説明:Yeast extract