Orders LinkContactUsLinkComponent
ニトリルグローブ、VWR®製
ニトリルグローブ、VWR®製
XL, Blue, 229 mm, 100/box
Catalog # 82026-430
Supplier:   VWR Collection
ニトリルグローブ、VWR®製
Catalog # 82026-430
Supplier:  VWR Collection
ニトリルグローブ、VWR®製
XL, Blue, 229 mm, 100/box
Price
$14.53
Log in for availability
1 * 100 items
Product Details & Documents
These ambidextrous examination gloves are ideal for use in industrial and laboratory applications as they protect against biohazards and a broad range of chemicals. They are made from a premium compounded nitrile for high manufacturing quality measures and product consistency.
  • Non sterile, powder-free, textured finish
  • Extra soft and comfortable for every day use
  • Rolled, beaded cuff allows easy donning
  • Protect hands from a broad range of chemicals
  • 100% latex-free to eliminate adverse reactions associated with natural rubber latex proteins
Specifications
  • 手袋のサイズ:
    XL
  • Packed:
    100/box
  • Colour:
    Blue
  • 長さ:
    229 mm
  • Seq:
    05
  • Palm thickness:
    0,12 mm
  • Cat. No.:
    82026-430
  • Latex / latex-free:
    Latex-free
  • Cuff thickness:
    0,1 mm
  • Disposable / reusable:
    Disposable
  • Cuff style:
    Beaded
  • Powdered / powder-free:
    Powder-free
  • Finish:
    Textured
  • Ambidextrous / hand-specific:
    Ambidextrous
  • Finger thickness:
    0,14 mm