pHインジケータースティック、BAKER-pHIX
pH indicator sticks, BAKER-pHIX, 6,0 - 7,7, 0,3/0,4
Supplier: Avantor
Jump to:
Product Details & Documents
BAKER-pHIX indicator sticks provide fast, easy, accurate measurement of pH ranges in laboratory.
Universal range sticks have resolution of 1,0 pH unit. Intermediate range sticks have resolutions of 0,3 to 0,5 pH units and narrow range sticks offer resolutions of 0,2 to 0,4 pH units.
: Packaged in easy to use plastic boxes containing 100 sticks.
Certificates
Search by Lot NumberSpecifications
- pH gradation:0,3/0,4
- pH range:6,0 - 7,7
- 説明:pH indicator sticks, BAKER-pHIX
- Seq:17