Orders LinkContactUsLinkComponent
pHインジケータースティック、BAKER-pHIX
pH indicator sticks, BAKER-pHIX, 6,0 - 7,7, 0,3/0,4
Catalog # 4396-01
Supplier:   Avantor
pHインジケータースティック、BAKER-pHIX
Catalog # 4396-01
Supplier:  Avantor
pHインジケータースティック、BAKER-pHIX
pHインジケータースティック、BAKER-pHIX
pH indicator sticks, BAKER-pHIX, 6,0 - 7,7, 0,3/0,4
Price
Inquire For Price
Log in for availability
1 * 100 items
Product Details & Documents
BAKER-pHIX indicator sticks provide fast, easy, accurate measurement of pH ranges in laboratory.
Universal range sticks have resolution of 1,0 pH unit. Intermediate range sticks have resolutions of 0,3 to 0,5 pH units and narrow range sticks offer resolutions of 0,2 to 0,4 pH units.
 : Packaged in easy to use plastic boxes containing 100 sticks.
Specifications
  • pH gradation:
    0,3/0,4
  • pH range:
    6,0 - 7,7
  • 説明:
    pH indicator sticks, BAKER-pHIX
  • Seq:
    17