Order Entry
Japan
Orders LinkContactUsLinkComponent
 

再封可能なバッグ、ZIPPIT®(ジッピット

101,6 µm, 355×610 mm
Catalog # VWRU89005-316
Supplier:   VWR Collection
再封可能なバッグ、ZIPPIT®(ジッピット
Catalog # VWRU89005-316
Supplier:  VWR Collection
再封可能なバッグ、ZIPPIT®(ジッピット
再封可能なバッグ、ZIPPIT®(ジッピット

101,6 µm, 355×610 mm

Price
$331.61
Log in for availability
1 * 250 items
Product Details & Documents

Plain ZIPPIT® reclosable zipper bags

  • Economical
  • Can be printed with your logo or instructions
  • Including bar code

 : In easy-to-use DispenserBag® convenience packaging, designed to function like a 'tissue soft pack'. Pull one bag at a time from the center package opening or open the reclosable zipper at the top to remove.

 : *The product(s) on this page are outside the scope of the EU Directive 2019/904

Specifications
  • 寸法:
    355×610 mm
  • Seq:
    18
  • 厚さ:
    101,6 µm