Order Entry
Japan
Orders LinkContactUsLinkComponent
保存袋、UVガード

保存袋、UVガード

76,2 µm, 203×203 mm
Catalog # VWRU89005-328
Supplier:   VWR Collection
保存袋、UVガード
Catalog # VWRU89005-328
Supplier:  VWR Collection
保存袋、UVガード
保存袋、UVガード

76,2 µm, 203×203 mm

Price
$266.09
Log in for availability
1 * 1.000 items
Product Details & Documents

UV Guard® UV protection bag is the amber UV blocker protective bags with the Color Line™. It prevents the bag contents from deterioration.

  • Premium zipper opens quickly and easily
  • Amber color makes identification of light-sensitive contents easy

Complies with USP XXII and USP XXIII procedure 66,1 with light transmission not exceeding 10% at any wavelength from 290 to 450 nm.

Specifications
  • 寸法:
    203×203 mm
  • Seq:
    05
  • 厚さ:
    76,2 µm