VWR®, 漏斗、粉体
120 mm, 30 mm, 30 mm
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 3.3 glass.
- Good chemical and physical properties
- Very good thermal resistance
- Quick and easy to clean
Smooth internal walls. For transferring powders or liquids.
Specifications
- Funnel O-Ø:120 mm
- Seq:05
- Stem O-Ø:30 mm
- Stem length:30 mm