VWR®, 漏斗、粉体
50 mm, 20 mm, 30 mm
Borosilicate 3.3 glass.
- Good chemical and physical properties
- Very good thermal resistance
- Quick and easy to clean
Smooth internal walls. For transferring powders or liquids.
- Funnel O-Ø:50 mm
- Seq:01
- Stem O-Ø:20 mm
- Stem length:30 mm