VWR®, バイオハザード標本袋
Biohazard specimen bags, LDPE, 229×152,4 mm, —
VWRU11215-684
Designed for safe and secure transport of a specimen container and documentation, these 51 µm thick bags feature a pouch to keep the paperwork separate from the container. They have a self-sealing closure and are tamper evident.
Documents
- 説明:Biohazard specimen bags
- 長さ×幅:229×152,4 mm
- Material:LDPE
- Seq:02
- 厚さ:—