Order Entry
Japan
Orders LinkContactUsLinkComponent
 

再封可能なバッグ、ZIPPIT®(ジッピット

50,8 µm, 152×152 mm
Catalog # VWRU89005-284
Supplier:   VWR Collection
再封可能なバッグ、ZIPPIT®(ジッピット
Catalog # VWRU89005-284
Supplier:  VWR Collection
再封可能なバッグ、ZIPPIT®(ジッピット
再封可能なバッグ、ZIPPIT®(ジッピット

50,8 µm, 152×152 mm

Price
$58.47
Log in for availability
1 * 1.000 items
Product Details & Documents

Plain ZIPPIT® reclosable zipper bags

  • Economical
  • Can be printed with your logo or instructions
  • Including bar code

 : In easy-to-use DispenserBag® convenience packaging, designed to function like a 'tissue soft pack'. Pull one bag at a time from the center package opening or open the reclosable zipper at the top to remove.

 : *The product(s) on this page are outside the scope of the EU Directive 2019/904

Specifications
  • 寸法:
    152×152 mm
  • Seq:
    06
  • 厚さ:
    50,8 µm