Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, 廃棄用バッグ

VWR®, 廃棄用バッグ

Autoclavable bags, clear, 3 l, 400×250 mm
Catalog # 129-0579
Supplier:   VWR Collection
VWR®, 廃棄用バッグ
Catalog # 129-0579
Supplier:  VWR Collection
VWR®, 廃棄用バッグ
VWR®, 廃棄用バッグ

Autoclavable bags, clear, 3 l, 400×250 mm

Price
Inquire For Price
Log in for availability
1 * 500 items
Product Details & Documents

For safe disposal of laboratory waste such as pipette tips, disposable Petri dishes, sample tubes, etc. PP, 40 µm thick, clear, with or without 'Biohazard' printing.

  • Heat resistant up to 145 °C
  • Autoclavable
Specifications
  • 容量:
    3 l
  • 説明:
    Autoclavable bags, clear
  • 長さ×幅:
    400×250 mm
  • Seq:
    02