VWR®, 廃棄用バッグ
Autoclavable bags, clear, with 'Biohazard' printing, 12 l, 660×305 mm
Supplier: VWR Collection
Jump to:
Product Details & Documents
For safe disposal of laboratory waste such as pipette tips, disposable Petri dishes, sample tubes, etc. PP, 40 µm thick, clear, with or without 'Biohazard' printing.
- Heat resistant up to 145 °C
- Autoclavable
Specifications
- 容量:12 l
- 説明:Autoclavable bags, clear, with 'Biohazard' printing
- 長さ×幅:660×305 mm
- Seq:10



