VWR®, 廃棄用バッグ
Autoclavable bags, clear, 2 l, 300×200 mm
Supplier: VWR Collection
Jump to:
Product Details & Documents
For safe disposal of laboratory waste such as pipette tips, disposable Petri dishes, sample tubes, etc. PP, 40 µm thick, clear, with or without 'Biohazard' printing.
- Heat resistant up to 145 °C
- Autoclavable
Specifications
- 容量:2 l
- 説明:Autoclavable bags, clear
- 長さ×幅:300×200 mm
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



