マグネット式スターラーバー、Spinvane® (スピンベーン)
3 or 5 ml vials and conical tubes
Catalog # VWRU58949-272
Supplier: VWR Collection
Jump to:
Product Details & Documents
PTFE covered, Alnico V magnetic core. Spinvane® Triangle shaped magnetic stirring bars are designed for stirring in test tubes, centrifuge tubes and micro vials.
- The wings can also be trimmed to size if needed
- Autoclave safe
Specifications
- For:3 or 5 ml vials and conical tubes
- Seq:02