Order Entry
Japan
Orders LinkContactUsLinkComponent
再封可能なバッグ、ZIPPIT®(ジッピット

再封可能なバッグ、ZIPPIT®(ジッピット

50,8 µm, 76×127 mm
Catalog # VWRU89005-278
Supplier:   VWR Collection
再封可能なバッグ、ZIPPIT®(ジッピット
Catalog # VWRU89005-278
Supplier:  VWR Collection
再封可能なバッグ、ZIPPIT®(ジッピット
再封可能なバッグ、ZIPPIT®(ジッピット

50,8 µm, 76×127 mm

Price
$28.14
Log in for availability
1 * 1.000 items
Product Details & Documents

Plain ZIPPIT® reclosable zipper bags

  • Economical
  • Can be printed with your logo or instructions
  • Including bar code

包装: In easy-to-use DispenserBag® convenience packaging, designed to function like a 'tissue soft pack'. Pull one bag at a time from the center package opening or open the reclosable zipper at the top to remove.

注意: *The product(s) on this page are outside the scope of the EU Directive 2019/904

Specifications
  • 寸法:
    76×127 mm
  • Seq:
    03
  • 厚さ:
    50,8 µm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.