Orders LinkContactUsLinkComponent
メスシリンダー、トールフォーム、PP
メスシリンダー、トールフォーム、PP
25 ml, 0,5 ml
Catalog # 612-3856
Supplier:   VWR Collection
メスシリンダー、トールフォーム、PP
Catalog # 612-3856
Supplier:  VWR Collection
メスシリンダー、トールフォーム、PP
メスシリンダー、トールフォーム、PP
25 ml, 0,5 ml
Price
$9.74
Log in for availability
1 * 1 items
Product Details & Documents
The 'nonwetting' polypropylene (PP) plastic surface of these single scale graduated cylinders eliminates formation of a concave meniscus and makes reading easier and more accurate. Chemically resistant, they are autoclavable at 121 °C (250 °F).
  • Single metric scale has raised numerals for easy reading
  • Flared top rims and large spouts facilitate pouring
  • Octagonal-shaped bases prevent tipping and rolling
Specifications
  • 容量:
    25 ml
  • Division:
    0,5 ml