Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, 真空デシケーター

VWR®, 真空デシケーター

With stopcock, 150 DN, 215×250 mm, 143 mm
  467-0087
VWR®, 真空デシケーター
  467-0087
VWR®, 真空デシケーター
VWR®, 真空デシケーター

With stopcock, 150 DN, 215×250 mm, 143 mm

 
 
   
1 * 1 items
 

Borosilicate 3.3 glass.

  • Precisely ground flat flanges and interchangeable tubulated lids
  • Standard ground joint NS 24/29 junction tube in the lid, complete with stopcock plus PTFE spindle
  • Base and lid with flat flange ground to fit
  • High mechanical, thermal and chemical resistance

Due to the increased wall thickness, these desiccators can be used up to the maximum vacuum that is technically possible.

Certifications: DIN 12491

 
  • 内径:
    143 mm
  • Nominal size:
    150 DN
  • Type:
    With stopcock
  • Seq:
    02
  • Ø ext.×H:
    215×250 mm