Orders LinkContactUsLinkComponent
VWR®, 秤量ボート
VWR®, 秤量ボート
64 mm, 29 mm
Catalog # 611-2388
Supplier:   VWR Collection
VWR®, 秤量ボート
Catalog # 611-2388
Supplier:  VWR Collection
VWR®, 秤量ボート
VWR®, 秤量ボート
64 mm, 29 mm
Price
Inquire For Price
Log in for availability
1 * 3 items
Product Details & Documents
Porcelain, glazed.
Documents
Specifications
  • 幅:
    29 mm
  • 長さ:
    64 mm
  • Seq:
    03