スクープ、セパレートハンドル付き、VWR®
Scoop, 165 mm
Catalog # VWRU82027-498
Supplier: VWR Collection
Jump to:
Product Details & Documents
Stainless steel.
- Sturdy stainless steel construction
- Handle and scoop are sold separately
The rounded tip of this scoop makes it easy to push into powders or break up solidified chemicals into smaller pieces, while its concave shape makes it ideal for picking them up.
Specifications
- 説明:Scoop
- 長さ:165 mm
- Seq:01