VWR®, シューズカバー、CPE
Blue, U*
Supplier: VWR Collection
Jump to:
Product Details & Documents
Basic shoe covers manufactured from a low-density chlorinated polyethylene fabric, which is designed to protect against dirt, grime, and liquid splash.
- CPE is water resistant and provides good resistance against chemicals
- Economical and lint-free
Ideal for laboratory use.
* One size fits all
Specifications
- 靴のサイズ:U*
- Colour:Blue
- Seq:03
- Cat. No.:113-1492
- Garment application:Laboratory