Orders LinkContactUsLinkComponent
スライドホルダ、VWR®製
Single slide holders, Blue, PP, 1
Catalog # VWRU95042-208
Supplier:   VWR Collection
スライドホルダ、VWR®製
Catalog # VWRU95042-208
Supplier:  VWR Collection
スライドホルダ、VWR®製
スライドホルダ、VWR®製
Single slide holders, Blue, PP, 1
Price
$88.83
Log in for availability
1 * 250 items
Product Details & Documents
Polypropylene resistant slide holders easy to open for durable protection.
 : FDA certified
 : 250 slides per box, four boxes per case.
Specifications
  • No. of slides:
    1
  • Colour:
    Blue
  • 説明:
    Single slide holders
  • Material:
    PP
  • Seq:
    01