スライドホルダ、VWR®製
Single slide holders, Blue, PP, 1
Catalog # VWRU95042-208
Supplier: VWR Collection
Jump to:
Product Details & Documents
Polypropylene resistant slide holders easy to open for durable protection.
: FDA certified
: 250 slides per box, four boxes per case.
Specifications
- No. of slides:1
- Colour:Blue
- 説明:Single slide holders
- Material:PP
- Seq:01