Order Entry
Japan
Orders LinkContactUsLinkComponent
 

VWR®, クリーンルーム用テープ、一般用途

Cleanroom tape, general purpose, Yellow, 25,4 mm
Catalog # 817-0094
Supplier:   VWR Collection
VWR®, クリーンルーム用テープ、一般用途
Catalog # 817-0094
Supplier:  VWR Collection
VWR®, クリーンルーム用テープ、一般用途
VWR®, クリーンルーム用テープ、一般用途

Cleanroom tape, general purpose, Yellow, 25,4 mm

Price
Inquire For Price
Log in for availability
1 * 1 Roll
Product Details & Documents

These multi-purpose vinyl tapes, wound on a plastic core, are ideal for use in cleanroom maintenance and support areas.

  • Ideal for colour coding, sealing, aisle marking and batching
  • Matt finish can be written on with a cleanroom pen

Length: 32,9 m.

Different colours available.

 : Single-packed in an easy tear, perforated bag.

Specifications
  • 幅:
    25,4 mm
  • Colour:
    Yellow
  • 説明:
    Cleanroom tape, general purpose
  • Seq:
    05