Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, クリーンルーム用テープ、一般用途

VWR®, クリーンルーム用テープ、一般用途

Cleanroom tape, general purpose, Dark blue, 25,4 mm
Catalog # 817-0087
Supplier:   VWR Collection
VWR®, クリーンルーム用テープ、一般用途
Catalog # 817-0087
Supplier:  VWR Collection
VWR®, クリーンルーム用テープ、一般用途
VWR®, クリーンルーム用テープ、一般用途

Cleanroom tape, general purpose, Dark blue, 25,4 mm

Price
Inquire For Price
Log in for availability
1 * 1 Roll
Product Details & Documents

These multi-purpose vinyl tapes, wound on a plastic core, are ideal for use in cleanroom maintenance and support areas.

  • Ideal for colour coding, sealing, aisle marking and batching
  • Matt finish can be written on with a cleanroom pen

Length: 32,9 m.

Different colours available.

包装: Single-packed in an easy tear, perforated bag.

Specifications
  • 幅:
    25,4 mm
  • Colour:
    Dark blue
  • 説明:
    Cleanroom tape, general purpose
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.