Order Entry
Japan
Orders LinkContactUsLinkComponent
 

VWR®, クリーンルーム用テープ、一般用途

Cleanroom tape, general purpose, Red, 25,4 mm
  817-0093
VWR®, クリーンルーム用テープ、一般用途
  817-0093
VWR®, クリーンルーム用テープ、一般用途
VWR®, クリーンルーム用テープ、一般用途

Cleanroom tape, general purpose, Red, 25,4 mm

 
 
   
1 * 1 Roll
 

These multi-purpose vinyl tapes, wound on a plastic core, are ideal for use in cleanroom maintenance and support areas.

  • Ideal for colour coding, sealing, aisle marking and batching
  • Matt finish can be written on with a cleanroom pen

Length: 32,9 m.

Different colours available.

 : Single-packed in an easy tear, perforated bag.

 
  • 幅:
    25,4 mm
  • Colour:
    Red
  • 説明:
    Cleanroom tape, general purpose
  • Seq:
    19