VWR®, デシケータープレート
Porcelain, 235 mm
467-0103
Porcelain desiccator plates. Central hole Ø 20 mm, filter holes Ø approximately 5 mm. Suitable for use with any desiccators with corresponding diameter.
: DIN 12911
- 直径:235 mm
- Type:Porcelain
- Seq:4
Porcelain desiccator plates. Central hole Ø 20 mm, filter holes Ø approximately 5 mm. Suitable for use with any desiccators with corresponding diameter.
: DIN 12911