VWR®, デシケータープレート
Porcelain, 190 mm
467-0102
Porcelain desiccator plates. Central hole Ø 20 mm, filter holes Ø approximately 5 mm. Suitable for use with any desiccators with corresponding diameter.
: DIN 12911
- 直径:190 mm
- Type:Porcelain
- Seq:3
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



