Order Entry
Japan
Orders LinkContactUsLinkComponent
 

VWR®, クリーンルーム用テープ、一般用途

Cleanroom tape, general purpose, Light green, 25,4 mm
Catalog # 817-0090
Supplier:   VWR Collection
VWR®, クリーンルーム用テープ、一般用途
Catalog # 817-0090
Supplier:  VWR Collection
VWR®, クリーンルーム用テープ、一般用途
VWR®, クリーンルーム用テープ、一般用途

Cleanroom tape, general purpose, Light green, 25,4 mm

Price
Inquire For Price
Log in for availability
1 * 1 Roll
Product Details & Documents

These multi-purpose vinyl tapes, wound on a plastic core, are ideal for use in cleanroom maintenance and support areas.

  • Ideal for colour coding, sealing, aisle marking and batching
  • Matt finish can be written on with a cleanroom pen

Length: 32,9 m.

Different colours available.

Packaging: Single-packed in an easy tear, perforated bag.

Specifications
  • 幅:
    25,4 mm
  • Colour:
    Light green
  • 説明:
    Cleanroom tape, general purpose
  • Seq:
    15
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.