Orders LinkContactUsLinkComponent
VWR®, メスフラスコ
1000 ml, 24/29, 0,40 ± ml
  612-3830
VWR®, メスフラスコ
  612-3830
VWR®, メスフラスコ
VWR®, メスフラスコ
1000 ml, 24/29, 0,40 ± ml
1 * 1 items
Borosilicate 3.3 glass, amber, class A.
  • White PE stopper
  • Marks and inscriptions in white enamel
  • Calibrated to contain (TC, In)
DIN EN ISO 1042
 : Batch number and batch certificate included.
  • NS:
    24/29
  • 容量:
    1000 ml
  • Seq:
    10
  • Tolerance:
    0,40 ± ml