VWR®, メスフラスコ
1000 ml, 24/29, 0,40 ± ml
Borosilicate 3.3 glass, amber, class A.
- White PE stopper
- Marks and inscriptions in white enamel
- Calibrated to contain (TC, In)
DIN EN ISO 1042
: Batch number and batch certificate included.
- NS:24/29
- 容量:1000 ml
- Seq:10
- Tolerance:0,40 ± ml