Orders LinkContactUsLinkComponent
VWR®, メスフラスコ
2000 ml, 29/32, 0,60 ± ml
Catalog # 612-3831
Supplier:   VWR Collection
VWR®, メスフラスコ
Catalog # 612-3831
Supplier:  VWR Collection
VWR®, メスフラスコ
VWR®, メスフラスコ
2000 ml, 29/32, 0,60 ± ml
Product Details & Documents
Borosilicate 3.3 glass, amber, class A.
  • White PE stopper
  • Marks and inscriptions in white enamel
  • Calibrated to contain (TC, In)
DIN EN ISO 1042
 : Batch number and batch certificate included.
Specifications
  • NS:
    29/32
  • 容量:
    2000 ml
  • Seq:
    11
  • Tolerance:
    0,60 ± ml