VWR®, メスフラスコ
2000 ml, 29/32, 0,60 ± ml
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 3.3 glass, amber, class A.
- White PE stopper
- Marks and inscriptions in white enamel
- Calibrated to contain (TC, In)
DIN EN ISO 1042
: Batch number and batch certificate included.
Specifications
- NS:29/32
- 容量:2000 ml
- Seq:11
- Tolerance:0,60 ± ml