Orders LinkContactUsLinkComponent
VWR®, メスフラスコ
VWR®, メスフラスコ
200 ml, 14/23, 0,15 ± ml
Catalog # 612-3827
Supplier:   VWR Collection
VWR®, メスフラスコ
Catalog # 612-3827
Supplier:  VWR Collection
VWR®, メスフラスコ
VWR®, メスフラスコ
200 ml, 14/23, 0,15 ± ml
Price
Inquire For Price
Log in for availability
1 * 2 items
Product Details & Documents
Borosilicate 3.3 glass, amber, class A.
  • White PE stopper
  • Marks and inscriptions in white enamel
  • Calibrated to contain (TC, In)
DIN EN ISO 1042
注文情報: Batch number and batch certificate included.
Specifications
  • NS:
    14/23
  • 容量:
    200 ml
  • Seq:
    07
  • Tolerance:
    0,15 ± ml