Orders LinkContactUsLinkComponent
VWR®, メスフラスコ
VWR®, メスフラスコ
25 ml, 10/19, 0,040 ± ml
Catalog # 612-3742
Supplier:   VWR Collection
VWR®, メスフラスコ
Catalog # 612-3742
Supplier:  VWR Collection
VWR®, メスフラスコ
VWR®, メスフラスコ
25 ml, 10/19, 0,040 ± ml
Price
Inquire For Price
Log in for availability
1 * 2 items
Product Details & Documents
Borosilicate 3.3 glass, class A.
  • PP white stopper
  • Marks and inscriptions in blue enamel
  • Calibrated to contain (TC, In)
DIN EN ISO 1042
Certifications: DIN EN ISO 1042
Ordering information: Batch number and batch certificate included.
Specifications
  • NS:
    10/19
  • 容量:
    25 ml
  • Seq:
    04
  • Tolerance:
    0,040 ± ml
  • Cat. No.:
    612-3742