VWR®, メスフラスコ
200 ml, 14/23, 0,150 ± ml
Borosilicate 3.3 glass, class A.
- PP white stopper
- Marks and inscriptions in blue enamel
- Calibrated to contain (TC, In)
DIN EN ISO 1042
: DIN EN ISO 1042
: Batch number and batch certificate included.
- NS:14/23
- 容量:200 ml
- Seq:07
- Tolerance:0,150 ± ml