Orders LinkContactUsLinkComponent
VWR®, メスフラスコ
500 ml, 19/26, 0,25 ± ml
  612-3829
VWR®, メスフラスコ
  612-3829
VWR®, メスフラスコ
VWR®, メスフラスコ
500 ml, 19/26, 0,25 ± ml
1 * 2 items
Borosilicate 3.3 glass, amber, class A.
  • White PE stopper
  • Marks and inscriptions in white enamel
  • Calibrated to contain (TC, In)
DIN EN ISO 1042
 : Batch number and batch certificate included.
  • NS:
    19/26
  • 容量:
    500 ml
  • Seq:
    09
  • Tolerance:
    0,25 ± ml