VWR®, メスフラスコ
25 ml, 10/19, 0,04 ± ml
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 3.3 glass, amber, class A.
- White PE stopper
- Marks and inscriptions in white enamel
- Calibrated to contain (TC, In)
DIN EN ISO 1042
: Batch number and batch certificate included.
Specifications
- NS:10/19
- 容量:25 ml
- Seq:04
- Tolerance:0,04 ± ml