Orders LinkContactUsLinkComponent
VWR®, メスフラスコ
25 ml, 10/19, 0,04 ± ml
Catalog # 612-3824
Supplier:   VWR Collection
VWR®, メスフラスコ
Catalog # 612-3824
Supplier:  VWR Collection
VWR®, メスフラスコ
VWR®, メスフラスコ
25 ml, 10/19, 0,04 ± ml
Price
Inquire For Price
Log in for availability
1 * 2 items
Product Details & Documents
Borosilicate 3.3 glass, amber, class A.
  • White PE stopper
  • Marks and inscriptions in white enamel
  • Calibrated to contain (TC, In)
DIN EN ISO 1042
 : Batch number and batch certificate included.
Specifications
  • NS:
    10/19
  • 容量:
    25 ml
  • Seq:
    04
  • Tolerance:
    0,04 ± ml