VWR®, メスフラスコ
50 ml, 12/21, 0,06 ± ml
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 3.3 glass, amber, class A.
- White PE stopper
- Marks and inscriptions in white enamel
- Calibrated to contain (TC, In)
DIN EN ISO 1042
注文情報: Batch number and batch certificate included.
Specifications
- NS:12/21
- 容量:50 ml
- Seq:05
- Tolerance:0,06 ± ml