VWR®, メスフラスコ
100 ml, 14/23, 0,10 ± ml
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 3.3 glass, amber, class A.
- White PE stopper
- Marks and inscriptions in white enamel
- Calibrated to contain (TC, In)
DIN EN ISO 1042
Ordering information: Batch number and batch certificate included.
Specifications
- NS:14/23
- 容量:100 ml
- Seq:06
- Tolerance:0,10 ± ml