VWR®, メスフラスコ
5 ml, 10/19, 0,040 ± ml
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 3.3 glass, class A.
- PP white stopper
- Marks and inscriptions in blue enamel
- Calibrated to contain (TC, In)
DIN EN ISO 1042
Certifications: DIN EN ISO 1042
Ordering information: Batch number and batch certificate included.
Documents
CoC VWR Volumetric Flask, Class A, NS Plastic Stopper.pdf COC_VWRI612-3738_03.19 COC_VWRI612-3738_GL0119 COC_VWRI612-3738_GL0122 coc_VWRI612-3738_GL0325
More
VWRI612-3738_TDS_EN VWRI612-3740_TDS_EN VWRI612-3744_TDS_EN VWRI612-3745_TDS_EN VWRI612-3820_TDS_EN
Datasheet
VWR® for Calibrations VWR® Glassware Solutions
Product Literature
Specifications
- NS:10/19
- 容量:5 ml
- Seq:01
- Tolerance:0,040 ± ml