VWR®, メスフラスコ
500 ml, 19/26, 0,250 ± ml
Borosilicate 3.3 glass, class A.
- PP white stopper
- Marks and inscriptions in blue enamel
- Calibrated to contain (TC, In)
DIN EN ISO 1042
: DIN EN ISO 1042
: Batch number and batch certificate included.
- NS:19/26
- 容量:500 ml
- Seq:09
- Tolerance:0,250 ± ml