ビーカー、低形状、PP
1000 ml, 100 - 1000 ml
Catalog # VWRU13890-148
Supplier: VWR Collection
Jump to:
Product Details & Documents
PP, translucent, graduated.
- Features no-drip spout for clean pouring
- Tapered walls for secure grip
- With an oval marking spot
Ideal for general laboratory procedures. These are easy-to-handle stackable beakers and is break-resistant, nontoxic, non-contaminating, and extremely chemical-resistant. Sturdy construction can withstand repeated autoclaving.
Specifications
- 目盛り範囲:100 - 1000 ml
- 容量:1000 ml
- Seq:06