VWR®, ワイプ、一般用
Tissue wipes, light duty, 1 ply, 213×114 mm, 280 wipes/box
Catalog # 82003-820
Supplier: VWR Collection
Jump to:
Product Details & Documents
Absorbent, soft, thick, low lint tissues made from virgin fibres. For light use in the laboratory, electronics, computer industry, pharmaceuticals, biotechnology and optics.
- High absorbency, in three formats, available in single or triple ply
- Practical dispenser box with anti-static, plastic film window for removal
- Table and wall-mounted dispensers for one handed removal of the wipes are available separately
Colour: White
Specifications
- Packed:280 wipes/box
- 説明:Tissue wipes, light duty, 1 ply
- 長さ×幅:213×114 mm



