VWR®, スライドメーラー
Slide mailers, with end opening, Assorted, PP, 43×24×88 mm, 5
Catalog # VWRU82003-431
Supplier: VWR Collection
Jump to:
Product Details & Documents
These boxes are designed for shipping, storing, or staining slides.
- Inside wall features slotted channels to keep slides separated and making removal easier
- Closes securely to keep slides safe - side opening
- Assorted colours (red, yellow, blue, purple, orange) for easier identification
Documents
VWR® Collection Selection Catalogue
Product Literature
Specifications
- No. of slides:5
- Colour:Assorted
- 説明:Slide mailers, with end opening
- Material:PP
- Seq:01
- W×D×H:43×24×88 mm