Orders LinkContactUsLinkComponent
VWR®, スライドメーラー
VWR®, スライドメーラー
Slide mailers, with end opening, Assorted, PP, 43×24×88 mm, 5
Catalog # VWRU82003-431
Supplier:   VWR Collection
VWR®, スライドメーラー
Catalog # VWRU82003-431
Supplier:  VWR Collection
VWR®, スライドメーラー
VWR®, スライドメーラー
Slide mailers, with end opening, Assorted, PP, 43×24×88 mm, 5
Price
$34.48
Log in for availability
1 * 25 items
Product Details & Documents
These boxes are designed for shipping, storing, or staining slides.
  • Inside wall features slotted channels to keep slides separated and making removal easier
  • Closes securely to keep slides safe - side opening
  • Assorted colours (red, yellow, blue, purple, orange) for easier identification
Specifications
  • No. of slides:
    5
  • Colour:
    Assorted
  • 説明:
    Slide mailers, with end opening
  • Material:
    PP
  • Seq:
    01
  • W×D×H:
    43×24×88 mm