解剖用鉗子、VWR®製
Straight, serrated, medium tips (blunt), 114 mm
Catalog # VWRU82027-388
Supplier: VWR Collection
Jump to:
Product Details & Documents
Dissecting forceps made of stainless steel. Available straight or curved and in different lengths.
Specifications
- Type:Straight, serrated, medium tips (blunt)
- 長さ:114 mm
- Seq:03