VWR® Basic Cleanroom Overalls, SMS
Without hood, Blue, 3XL
Catalog # 113-1095
Supplier: VWR Collection
Jump to:
Product Details & Documents
These cleanroom overalls are manufactured from a latex-free, multi-layer spun-bonded - meltblown - spun-bonded (SMS) fabric, that offers high tensile strength while remaining soft, comfortable and breathable.
- Tapered collar, inset sleeves, zip closure, elasticated cuffs
- Fluid and particulate barrier
- Light chemical splash resistance
Documents
Specifications
- 衣服のサイズ:3XL
- Colour:Blue
- 説明:Without hood
- Seq:12
- Cat. No.:113-1095
- Garment application:Cleanroom|Laboratory